missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome b5 Outer Mitochondrial Membrane Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97403-100ul
This item is not returnable.
View return policy
Description
Cytochrome b5 Outer Mitochondrial Membrane Polyclonal antibody specifically detects Cytochrome b5 Outer Mitochondrial Membrane in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Cytochrome b5 Outer Mitochondrial Membrane | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| CYB5M, CYB5-M, CYPB5M, Cytochrome b5 outer mitochondrial membrane isoform, cytochrome b5 type B, cytochrome b5 type B (outer mitochondrial membrane), DKFZp686M0619, OMB5, outer mitochondrial membrane cytochrome b5, type 2 cyt-b5 | |
| Produced in rabbits immunized with E. coli-derived Human Cytochrome b5 Outer Mitochondrial Membrane fragment. | |
| 100 μg | |
| Cancer, Endocrinology, Signal Transduction | |
| 80777 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction