missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome b reductase 1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £299.00
Specifications
| Antigen | Cytochrome b reductase 1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18318461
|
Novus Biologicals
NBP3-15534-20UL |
20 μg |
£139.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18394657
|
Novus Biologicals
NBP3-15534-100UL |
100 μg |
£299.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytochrome b reductase 1 Polyclonal antibody specifically detects Cytochrome b reductase 1 in Human samples. It is validated for Western BlotSpecifications
| Cytochrome b reductase 1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| cytochrome b reductase 1, DCYTBFRRS3FLJ23462, Duodenal cytochrome b, Ferric-chelate reductase 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 215-286 of human Cytochrome b reductase 1 (NP_079119.3). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Signal Transduction | |
| PBS, 50% glycerol, pH7.3 | |
| 79901 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title