missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
C17 is a cytokine-like protein specifically expressed in bone marrow and cord blood mononuclear cells that bear the CD34 surface marker (Liu et al., 2000 ).
Specifications
Specifications
| Antigen | CYTL1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.18 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CYTL1 |
| Gene Accession No. | Q9NRR1 |
| Gene Alias | C17, C4orf4, CYTL1, cytokine like 1, Cytokine like protein 1, Protein C17 |
| Gene Symbols | CYTL1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?