missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CYTL1 Polyclonal antibody specifically detects CYTL1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | CYTL1 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL |
| Formulation | PBS (pH 7.2) and 40% Glycerol |
| Gene Alias | C17, C4orf4cytokine-like protein C17, cytokine-like 1, cytokine-like protein 1, Protein C17 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: TCYSRMRALSQEITRDFNLLQVSEPSEPCVRYLPRLYLDIHNYCVLDKLRDFVAS |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?