missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CytKick™ Max Autosampler Upgrade Kit

Product Code. 17112534
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17112534 1 kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17112534 Supplier Invitrogen™ Supplier No. A43045
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Enables the upgrade of a CytKick Autosampler to a CytKick Max Autosampler

  • The CytKick Max Autosampler Upgrade Kit enables the upgrade of a CytKick Autosampler to a CytKick Max Autosampler.
  • The upgrade is carried out by either a field service engineer or field application specialist and provides the following features: faster plate times (22 minutes for a 96-well plate or 88 minutes for a 384-well plate) with a boost mode, user-definable customized plate definitions allowing the use of any ANSI spec plates, evaporation protection with foil covers, automated tube sample acquisition for 1.5- and 2.0-mL Eppendorf tubes, and passive cooling capability.
TRUSTED_SUSTAINABILITY

Specifications

Product Line CytKick
Quantity 1 kit
Shipping Condition Room Temperature
For Use With (Equipment) Attune Acoustic Focusing Cytometer
Product Type Autosampler Upgrade Kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.