Learn More
Invitrogen™ CytKick™ Autosampler Sample Syringe, 1 mL
Product Code. 17246336
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
Each
Product Code. Quantity Unit Size
17246336 1 mL Each
1 options
Product Code. 17246336 Supplier Invitrogen™ Supplier No. 100054593CST
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This 1-mL sample syringe is used with the fluidics system of the CytKick and CytKick Max autosampler to move sample from the autosampler to the flow cell of the Attune NxT and Attune CytPix flow cytometers.
This 1-mL sample syringe is used with the fluidics system of the CytKick and CytKick Max autosampler to move sample from the autosampler to the flow cell of the Attune NxT and Attune CytPix flow cytometers. Replacement of this syringe is recommended as part of instrument maintenance.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 mL
Capacity 1 mL
For Use With (Equipment) CytKick and CytKick Max Autosamplers
Product Type Autosampler Syringe