Learn More
Cytiva Cytiva Dissociation Enzyme Mix C - Brain
Product Code. 31703032 Shop All Cytiva Products
Change view
Click to view available options
Form:
Lyophilized
Unit Size:
1 set
Product Code. Form Unit Size
31703032 Lyophilized 1 set
1 options
Product Code. 31703032 Supplier Cytiva Supplier No. 29740240
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Enzymatic mixes for brain tissue optimized for manual and semi-automated dissociation in single-cell workflows.
  • Dissociation enzyme mixes (for research only) for brain are kits intended for the dissociation of brain tissues in single-cell sample preparation workflows
  • The kits have been optimized for use with manual and semi-automated tissue disaggregation techniques
  • Work effectively at either 4°C or 37°C
  • Improved single cell viability and reduced cellular aggregation
  • Ready-made solution for brain tissue - Optimized for the dissociation of the different segments that are operative in respective tissue
  • Optimized for use with manual tissue disaggregation techniques and semi-automated techniques including the VIA Extractor™ tissue disaggregator
  • Performance data is not available for Enzyme mix C (for brain) using semi-automated methods at temperatures of 37°C.
TRUSTED_SUSTAINABILITY
Specifications
Form Lyophilized
Product Type Cell Culture Dissociation Reagent
Sterility Non-sterile
With Additives None
Without Additives None