Learn More
Cytiva Cytiva Dissociation Enzyme Mix A - Lung
Product Code. 31703018 Shop All Cytiva Products
Change view
Click to view available options
Form:
Lyophilized
Unit Size:
1 set
Product Code. Form Unit Size
31703018 Lyophilized 1 set
1 options
Product Code. 31703018 Supplier Cytiva Supplier No. 29740243
Focus on Regenerative Therapies
Use promo code 28027
View all promotions
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Enzymatic mixes for lung tissue optimized for manual and semi-automated dissociation in single-cell workflows.
  • Dissociation enzyme mixes (for research only) for lung are kits intended for the dissociation of lung tissues in single-cell sample preparation workflows
  • The kits have been optimized for use with manual and semi-automated tissue disaggregation techniques
  • Work effectively at either 4°C or 37°C
  • Improved single cell viability and reduced cellular aggregation
  • Ready-made solution for lung tissue - Optimized for the dissociation of the different segments that are operative in respective tissue
  • Optimized for use with manual tissue disaggregation techniques and semi-automated techniques including the VIA Extractor™ tissue disaggregator
  • Performance data is not available for Enzyme mix A (for lung) using semi-automated methods at temperatures of 37°C
TRUSTED_SUSTAINABILITY
Specifications
Form Lyophilized
Product Type Cell Culture Dissociation Reagent
Sterility Non-sterile
With Additives None
Without Additives None