missing translation for 'onlineSavingsMsg'
Learn More

Cytisinicline, MedChemExpress

Product Code. 16413846
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16413846 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16413846 Supplier Medchem Express Supplier No. HYN0175

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cytisinicline (Cytisine) is an alkaloid that occurs naturally in several plant genera, such as Laburnum and Cytisus. Cytisinicline (Cytisine) is a partial agonist of α4β2 nAChRs, and partial to full agonist at β4 containing receptors and α7 receptors. Has been used medically to help with smoking cessation.

Cytisinicline (Cytisine) is an alkaloid that occurs naturally in several plant genera, such as Laburnum and Cytisus. Cytisinicline (Cytisine) is a partial agonist of α4β2 nAChRs, and partial to full agonist at β4 containing receptors and α7 receptors. Has been used medically to help with smoking cessation.

Specifications

Chemical Name or Material Cytisinicline
CAS 485-35-8
Color White
Purity Grade Notes Research
Molecular Formula C11H14N2O
Quantity 25 mg
Synonym Cytisine Sophorine Baptitoxine
Solubility Information DMSO : 125 mg/mL (657.06 mM; Need ultrasonic)
SMILES O=C1N(C[C@@]2([H])C[C@]3([H])CNC2)C3=CC=C1
Molecular Weight (g/mol) 190.24
Formula Weight 190.24
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.