missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cytidine, 99+%

Product Code. 10080860 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10080860 10 g 10g
10182420 50 g 50g
10629094 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10080860 Supplier Thermo Scientific Chemicals Supplier No. 111810100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 65-46-3 | C9H13N3O5 | 243.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-46-3
Molecular Formula C9H13N3O5
Molecular Weight (g/mol) 243.22
MDL Number MFCD00006545
InChI Key UHDGCWIWMRVCDJ-UHFFFAOYNA-N
Synonym cytidine, cytosine riboside, 1-beta-d-ribofuranosylcytosine, 1beta-ribofuranosylcytosine, 4-amino-1-beta-d-ribofuranosyl-2 1h-pyrimidinone, beta-d-ribofuranoside, cytosine-1, 1-beta-ribofuranosylcytosine, cytidin, zytidin, 4-amino-1beta-d-ribofuranosyl-2 1h-pyrimidinone
PubChem CID 6175
ChEBI CHEBI:17562
IUPAC Name 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1)C1OC(CO)C(O)C1O

Specifications

Absorbance 0.43 to 0.47 (A250/A260), 2.05 to 2.15 (A280/A260)
Melting Point 210.0°C to 217.0°C
Color White
Loss on Drying 0.5% max. (105°C, 5 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Beilstein 31,24
Merck Index 15,2783
Specific Rotation + 31.50
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acids and alkalies,insoluble in organic solvents
Spectrophotometry 98 to 102% (lambda max. 280nm,pH 1) molar absorptivity 13000+/-2%,on dry basis
Transmittance 98% min. (c=1, H2O, 430nm) 1cm cell
Formula Weight 243.22
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Cytidine, 99.5%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.