missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cytidine 5'-triphosphate, disodium salt hydrate, 95%

Product Code. 10062840
Click to view available options
500mg; Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 215.0°C to 218.0°C
CAS 652154-13-7
Color White
Physical Form Crystalline Powder
Quantity 500 mg
Assay Percent Range 94% min. (HPLC)
Molecular Formula C9H18N3Na2O16P3
MDL Number MFCD00150807
Synonym cytidine 5'-triphosphate sodium salt, cytidine 5'-triphosphate tetrasodium
Solubility Information Solubility in water: 200g/L (20°C)
InChI Key KNSYTSUGTLJHPS-ODQFIEKDSA-L
SMILES O.O.[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]1O
IUPAC Name [[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphono hydrogen phosphate;sodium
Molecular Weight (g/mol) 563.15
PubChem CID 129893565
Formula Weight 527.12
Percent Purity 95%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Cytidine 5′-triphosphate,disodium salt hydrate, 95%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'