missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Protein-A/G Protein

Product Code. p-7151046
Change view
Click to view available options
Quantity:
1 mg
10 mg
100 mg
Unit Size:
100mg
10mg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15932699 1 mg 1mg
15922699 10 mg 10mg
15912699 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15932699 Supplier enQuireBio™ Supplier No. QP131631mg
Add to basket
Add to basket
This item is not returnable. View return policy

A cDNA sequence encoding the Protein-A/G was constructed and used to recombinantly synthesize the protein.

Specifications

Name Protein-A/G Protein
Quantity 1 mg
Regulatory Status RUO
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Other
Species E. coli
Protein Tag His
Sequence MNAAQHDEAQ QNAFYQVLNM PNLNADQRNG FIQSLKDDPS QSANVLGEAQ KLNDSQAPKA DAQQNNFNKD QQSAFYEILN MPNLNEAQRN GFIQSLKDDP SQSTNVLGEA KKLNESQAPK ADNNFNKEQQ NAFYEILNMP NLNEEQRNGF IQSLKDDPSQ SANLLSEAKK LNESQAPKAD NKFNKEQQNA FYEILHLPNL NEEQRNGFIQ SLKDDPSQSA NLLAEAKKLN DAQAPKADNK FNKEQQNAFY EILHLPNLTE EQRNGFIQSL KDDPSVSKEI LAEAKKLNDA QAPKEEDSLE GSGSGTYKLI LNGKTLKGET TTEAVDAATA EKVFKQYAND NGVDGEWTYD DATKTFTVTE KPEVIDASEL TPAVTTYKLV INGKTLKGET TTEAVDAATA EKVFKQYAND NGVDGEWTYD DATKTFTVTE KPEVIDASEL TPAVTTYKLV INGKTLKGET TTKAVDAETA EKAFKQYAND NGVDGVWTYD DATKTFTVTE KLAAALEHHH HHH
Buffer Protein-A/G was lyophilized without any additives.
Purity or Quality Grade Greater than 96.0% as determined by: (a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.