missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cytidine-5'-monophosphate disodium salt, 99+%

Product Code. 15405629
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15405629 5 g 5g
15495619 1 g 1g
15415629 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15405629 Supplier Thermo Scientific Chemicals Supplier No. J63376.06
  • ONLINE EXCLUSIVE PRICE
    £61.50
    £55.65 / 5g
    Save £5.85
    9% Off
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6757-06-8 | C9H12N3Na2O8P | 367.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6757-06-8
Molecular Formula C9H12N3Na2O8P
Molecular Weight (g/mol) 367.16
MDL Number MFCD00150830
InChI Key INTPYBRGLGSMRA-IJRYJGAHNA-L
Synonym cytidine 5'-monophosphate disodium salt, disodium cmp, cmp disodium salt, cytidine-5'-monophosphate disodium salt, disodium 5'-cmp, 5'-cmp disodium salt, disodium 5'-o-phosphonatocytidine, 5'-cytidylic acid, disodium salt, disodium cytidine 5'-monophosphate, cytidine monophosphate disodium salt
PubChem CID 94332
IUPAC Name disodium;[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
SMILES [Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]1O

Specifications

Color White
Quantity 5 g
Formula Weight 367.16
Percent Purity ≥99%
Physical Form Crystalline Powder
Chemical Name or Material Cytidine-5'-monophosphate disodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.