missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cytidine 5'-monophosphate, 97%, from yeast

Product Code. 13418668 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13418668 5 g Glass bottle 5g
13408668 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13418668 Supplier Thermo Scientific Chemicals Supplier No. 449490050
 Restricted Item
Estimated Shipment: 20-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 63-37-6 | C9H14N3O8P | 323.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-37-6
Molecular Formula C9H14N3O8P
Molecular Weight (g/mol) 323.20
MDL Number MFCD00006544
InChI Key IERHLVCPSMICTF-NHBCCFCANA-N
Synonym 5'-cytidylic acid, cytidylic acid, cytidine 5'-monophosphate, cytidine monophosphate, cytidine-5'-monophosphate, 5'-cmp, cytidylate, cytidine 5'-phosphate, cmp nucleotide, cytidine 5'-phosphoric acid
PubChem CID 6131
ChEBI CHEBI:17361
IUPAC Name [(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate
SMILES NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP(O)(O)=O)[C@@H](O)[C@H]1O

Specifications

Melting Point 222°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: soluble
Formula Weight 323.2
Percent Purity 97%
Chemical Name or Material Cytidine 5'-monophosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.