missing translation for 'onlineSavingsMsg'
Learn More

Cytidine 5'-Diphosphocholine, TRC

Product Code. 30137477
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137477 2.5 g 2.5g
30012368 500 mg 500mg
30005382 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137477 Supplier Toronto Research Chemicals Supplier No. TRCC9983132.5G
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 987-78-0
Molecular Formula C14 H26 N4 O11 P2
Molecular Weight (g/mol) 488.32
Synonym Cytidine 5'-(trihydrogen diphosphate), P'-[2-(trimethylammonio)ethyl] ester, inner salt, Choline, hydroxide, 5'-ester with cytidine 5'-(trihydrogen pyrophosphate), inner salt (8CI), Cytidine 5'-(trihydrogen diphosphate), P'-[2-(trimethylammonio)ethyl] ester, hydroxide, inner salt, Audes, CDP-choline, Cereb, Choline 5'-cytidine diphosphate, Choline cytidine diphosphate, Citicholine, Citicoline, Citidoline, Citifar, Cognizin, Colite, Corenalin, Cyscholin, Cytidine 5'-(choline diphosphate), Cytidine 5'-(cholinyl pyrophosphate), Cytidine 5'-diphosphate choline, Cytidine 5'-diphosphocholine, Cytidine choline diphosphate, Cytidine diphosphate choline, Cytidine diphosphate choline ester, Cytidine diphosphocholine, Cytidine diphosphorylcholine, Cytidoline, Difosfocin, Emicholine F, Ensign, Haocolin, Hornbest, NSC 122002, Neucolis, Nicholin, Nicolin, Niticolin, Reagin, Recofnan, Recognan, Rexort, Sintoclar, Somazina, Somazine, Suncholin
IUPAC Name [[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] 2-(trimethylazaniumyl)ethyl phosphate
SMILES C[N+](C)(C)CCOP(=O)([O-])OP(=O)(O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=CC(=NC2=O)N

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C14H26N4O11P2/c1-18(2,3)6-7-26-30(22,23)29-31(24,25)27-8-9-11(19)12(20)13(28-9)17-5-4-10(15)16-14(17)21/h4-5,9,11-13,19-20H,6-8H2,1-3H3,(H3-,15,16,21,22,23,24,25)/t9-,11-,12-,13-/m1/s1
Formula Weight 488.1073
Chemical Name or Material Cytidine-5'-diphosphocholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.