missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Cytarabine
Click to view available options
Quantity:
50 mg
Unit Size:
10mg
Description
Nucleoside analog of deoxycytidine; inhibits DNA replication by incorporating into DNA (IC50 = 0.04 μM in L1210 and CEM cell lines). Displays no inhibitory effects on RNA synthesis. Causes S phase cell cycle arrest in ML-1 cell lines; cytotoxic in L5817Y leukemia cells. Antineoplastic and antileukemic agent.
Specifications
Specifications
| CAS | 147-94-4 |
| Quantity | 50 mg |
| Molecular Formula | C9H13N3O5 |
| Formula Weight | 243.22 |
| Purity | >99% |
| Molecular Weight (g/mol) | 243.219 |
| Chemical Name or Material | Cytarabine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction