missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Cytarabine hydrochloride

Product Code. 16456246
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16456246 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16456246 Supplier MedChemExpress Supplier No. HY13605A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cytarabine hydrochloride, a nucleoside analog, causes S phase cell cycle arrest and inhibits DNA polymerase. Cytarabine inhibits DNA synthesis with an IC50 of 16 nM. Cytarabine hydrochloride has antiviral effects against HSV.

Specifications

Molecular Weight (g/mol) 279.68 g/mol
Product Type Reference compound
Content And Storage 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Shipping Condition Room temperature
Purity 95.0%
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (178.78 mM∣Need ultrasonic)
For Use With (Application) Cancer-Kinase/protease
CAS 69-74-9
Synonym Cytosine β-D-arabinofuranoside hydrochloride∣ Cytosine Arabinoside hydrochloride∣ Ara-C hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.