missing translation for 'onlineSavingsMsg'
Learn More

Cysteinyl Leukotriene R2/CysLTR2 Antibody, Novus Biologicals™

Product Code. 18386612
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18386612 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18386612 Supplier Bio-Techne Supplier No. NBP314292

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Cysteinyl Leukotriene R2/CysLTR2 Polyclonal antibody specifically detects Cysteinyl Leukotriene R2/CysLTR2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cysteinyl Leukotriene R2/CysLTR2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias CysLT(2), CYSLT2KPG_011, CYSLT2RHG57, CysLTR2, cysteinyl leukotriene CysLT2 receptor, cysteinyl leukotriene receptor 2, G protein-coupled receptor, G-protein coupled receptor GPCR21, G-protein coupled receptor HG57, hGPCR21, HPN321
Host Species Rabbit
Immunogen Synthetic 20 amino acid peptide from C-Terminus cytoplasmic domain of human CYSLTR2.
Purification Method Immunogen affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 57105
Target Species Human
Content And Storage Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Product Type Antibody
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.