missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cysteinyl Leukotriene R2/CysLTR2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £484.00
Specifications
| Antigen | Cysteinyl Leukotriene R2/CysLTR2 |
|---|---|
| Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18487781
|
Novus Biologicals
NBP2-13897 |
0.1 mL |
£484.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18484512
|
Novus Biologicals
NBP2-13897-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cysteinyl Leukotriene R2/CysLTR2 Polyclonal antibody specifically detects Cysteinyl Leukotriene R2/CysLTR2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Cysteinyl Leukotriene R2/CysLTR2 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| GPCR | |
| PBS (pH 7.2) and 40% Glycerol | |
| 57105 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CysLT(2), CYSLT2KPG_011, CYSLT2RHG57, CysLTR2, cysteinyl leukotriene CysLT2 receptor, cysteinyl leukotriene receptor 2, G protein-coupled receptor, G-protein coupled receptor GPCR21, G-protein coupled receptor HG57, hGPCR21, HPN321 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: KFMSLQPSISVSEMEPNGTFSNNNSRNCTIE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title