missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cysteinyl Leukotriene R2/CysLTR2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92327-0.1ml
This item is not returnable.
View return policy
Description
Cysteinyl Leukotriene R2/CysLTR2 Polyclonal antibody specifically detects Cysteinyl Leukotriene R2/CysLTR2 in Mouse samples. It is validated for Western Blot
Specifications
| Cysteinyl Leukotriene R2/CysLTR2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CysLT(2), CYSLT2KPG_011, CYSLT2RHG57, CysLTR2, cysteinyl leukotriene CysLT2 receptor, cysteinyl leukotriene receptor 2, G protein-coupled receptor, G-protein coupled receptor GPCR21, G-protein coupled receptor HG57, hGPCR21, HPN321 | |
| A synthetic peptide corresponding to a sequence within amino acids 200 to the C-terminus of human CYSLTR2 (NP_001295394.1). TMNYIALVVGCLLPFFTLSICYLLIIRVLLKVEVPESGLRVSHRKALTTIIITLIIFFLCFLPYHTLRTVHLTTWKVGLCKDRLHKALVITLALAAANACFNPLLYYFAGENFKDRLKSALRKGHPQKAKTKCVFPVSVWLRKETRV | |
| 0.1 mL | |
| GPCR | |
| 57105 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction