missing translation for 'onlineSavingsMsg'
Learn More

Cysteine Conjugate beta-Lyase/CCBL1 Antibody [PerCP], Novus Biologicals™

Product Code. 30738081 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30738081 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30738081 Supplier Novus Biologicals Supplier No. AF7817C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Cysteine Conjugate beta-Lyase/CCBL1 Polyclonal antibody specifically detects Cysteine Conjugate beta-Lyase/CCBL1 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cysteine Conjugate beta-Lyase/CCBL1
Applications Western Blot
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias cysteine conjugate-beta lyase, cytoplasmic, cysteine conjugate-beta lyase; cytoplasmic (glutamine transaminase K, Cysteine-S-conjugate beta-lyase, EC 2.6.1.64, EC 2.6.1.7, EC 4.4.1.13, FLJ95217, Glutamine transaminase K, glutamine-phenylpyruvate aminotransferase, Glutamine--phenylpyruvate transaminase, GTKMGC29624, KAT1, KATIbeta-lysase, kidney, kyneurenine aminotransferase, kyneurenine aminotransferase), Kynurenine aminotransferase I, kynurenine--oxoglutarate transaminase 1, Kynurenine--oxoglutarate transaminase I
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Cysteine Conjugate β-Lyase/CCBL1, Thr182-Leu422, Accession # Q16773
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 883
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.