missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cysteine Conjugate beta-Lyase/CCBL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£342.00 - £470.00
Specifications
| Antigen | Cysteine Conjugate beta-Lyase/CCBL1 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18456970
|
Novus Biologicals
NBP1-83315-25ul |
25 μL |
£342.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18294217
|
Novus Biologicals
NBP1-83315 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cysteine Conjugate beta-Lyase/CCBL1 Polyclonal specifically detects Cysteine Conjugate beta-Lyase/CCBL1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Cysteine Conjugate beta-Lyase/CCBL1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 883 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:MAKQLQARRLDGIDYNPWVEFVKLASEHDVVNLGQGFPDFPPPDFAVEAFQHAVSGDFMLNQYTKTFGYPPLTKILASFFGELLGQEIDPLRNVLVTVGGYG | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| cysteine conjugate-beta lyase, cytoplasmic, cysteine conjugate-beta lyase; cytoplasmic (glutamine transaminase K, Cysteine-S-conjugate beta-lyase, EC 2.6.1.64, EC 2.6.1.7, EC 4.4.1.13, FLJ95217, Glutamine transaminase K, glutamine-phenylpyruvate aminotransferase, Glutamine--phenylpyruvate transaminase, GTKMGC29624, KAT1, KATIbeta-lysase, kidney, kyneurenine aminotransferase, kyneurenine aminotransferase), Kynurenine aminotransferase I, kynurenine--oxoglutarate transaminase 1, Kynurenine--oxoglutarate transaminase I | |
| CCBL1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title