missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Cystatin SN Monoclonal antibody specifically detects Cystatin SN in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Cystatin SN |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | MM0234-5N51 |
| Conjugate | PerCP |
| Dilution | Western Blot |
| Formulation | PBS |
| Gene Alias | cystain-SA-I, cystatin 1, cystatin SA-I, cystatin SN, cystatin-1, cystatin-SN, Salivary cystatin-SA-1, type 2 family |
| Host Species | Mouse |
| Immunogen | Human recombinant Cystatin SN |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?