missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin SA Antibody (OTI1D6), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70466H
This item is not returnable.
View return policy
Description
Cystatin SA Monoclonal specifically detects Cystatin SA in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Cystatin SA | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| CST2 | |
| Full length recombinant protein of human Cystatin SA (NP_001313) produced in HEK293T cells. | |
| 0.1 mL | |
| 1470.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI1D6 | |
| Western Blot, Immunohistochemistry | |
| cystatin 2, cystatin S5, cystatin SA, Cystatin-2, Cystatin-S5, cystatin-SA, cysteine-proteinase inhibitor, MGC71924, salivary cysteine (thiol) protease inhibitor | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction