missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin SA Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92665-0.02ml
This item is not returnable.
View return policy
Description
Cystatin SA Polyclonal antibody specifically detects Cystatin SA in Human, Mouse samples. It is validated for Western Blot
Specifications
| Cystatin SA | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| cystatin 2, cystatin S5, cystatin SA, Cystatin-2, Cystatin-S5, cystatin-SA, cysteine-proteinase inhibitor, MGC71924, salivary cysteine (thiol) protease inhibitor | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-141 of human CST2 (NP_001313.1). AWSPQEEDRIIEGGIYDADLNDERVQRALHFVISEYNKATEDEYYRRLLRVLRAREQIVGGVNYFFDIEVGRTICTKSQPNLDTCAFHEQPELQKKQLCSFQIYEVPWEDRMSLVNSRCQEA | |
| 0.02 mL | |
| Cell Biology | |
| 1470 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction