missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin SA Antibody (016), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89917AF405
This item is not returnable.
View return policy
Description
Cystatin SA Monoclonal specifically detects Cystatin SA in Human samples. It is validated for ELISA, Sandwich ELISA Detection.
Specifications
| Cystatin SA | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 1470 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 16 | |
| ELISA, Sandwich ELISA Detection | |
| cystatin 2, cystatin S5, cystatin SA, Cystatin-2, Cystatin-S5, cystatin-SA, cysteine-proteinase inhibitor, MGC71924, salivary cysteine (thiol) protease inhibitor | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Cystatin SA (NP_001313.1; Met 1-Ala 141). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction