missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Cystatin S Antibody Pair [HRP]

Product Code. 18259907 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18259907

Brand: Novus Biologicals™ NBP27962315PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Cynomolgus Monkey Cystatin S.
TRUSTED_SUSTAINABILITY

Specifikationer

Assay Range 62.5 pg/mL
Assay Sensitivity 62.5-4000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Cynomolgus Monkey Cystatin S.
Target Species Cynomolgus Monkey
For Use With (Application) Sandwich ELISA
Gene Alias cystatin 4, cystatin S, Cystatin-4, cystatin-S, cystatin-SA-III, MGC71923, Salivary acidic protein 1
Gene ID (Entrez) 1472
Gene Symbol CST4
Host Species Mouse
Kit Contents
  • Detection Antibody: 0.5 mg/mL mouse anti-Cystatin S monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Capture Antibody: 0.1 mg/mL of mouse anti-Cystatin S monoclonal antibody.
  • Standard: Each vial contains 30 ng of recombinant Cystatin S.
  • Quantity 15 Plates
    Regulatory Status RUO
    Storage Requirements Storage is content dependent.
    Target Cystatin S
    Technique 1. Capture Antibody: Dilute to a working concentration of 2.0 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 1 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute standard powder with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 4000 pg/mL is recommended.
    Visa mer Visa mindre
    Korrigering av produktinnehåll

    Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

    Produkttitel

    Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

    Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.