missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin F Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98950AF647
This item is not returnable.
View return policy
Description
Cystatin F Polyclonal specifically detects Cystatin F in Human samples. It is validated for ELISA.
Specifications
| Cystatin F | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 8530 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| Alexa Fluor 647 | |
| CMAP, cystatin F (leukocystatin), Cystatin-7, cystatin-F, Cystatin-like metastasis-associated protein, leukocystatin | |
| Produced in rabbits immunized with purified, recombinant Cystatin F (NP_003641.2; Met 1-His 145). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction