missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin E/M/CST6 Rabbit anti-Human, Clone: 34, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89500-100UL
This item is not returnable.
View return policy
Description
Cystatin E/M/CST6 Monoclonal specifically detects Cystatin E/M/CST6 in Human samples. It is validated for ELISA.
Specifications
| Cystatin E/M/CST6 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 34 | |
| ELISA 1:5000-1:10000 | |
| cystatin 6, cystatin E/M, cystatin M, cystatin M/E, Cystatin-6, Cystatin-E, cystatin-M, cysteine proteinase inhibitor | |
| Recombinant Human Cystatin E/M/CST6 protein | |
| 100 μg | |
| Cancer | |
| 1474 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction