missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin E/M/CST6 Antibody (RM0217-6F49), mFluor Violet 450 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Cystatin E/M/CST6 Monoclonal antibody specifically detects Cystatin E/M/CST6 in Mouse samples. It is validated for Western Blot, Neutralization
Specifications
Specifications
| Antigen | Cystatin E/M/CST6 |
| Applications | Western Blot, Neutralization |
| Classification | Monoclonal |
| Clone | RM0217-6F49 |
| Conjugate | mFluor Violet 450 SE |
| Dilution | Western Blot, Block/Neutralize |
| Formulation | 50mM Sodium Borate |
| Gene Alias | cystatin 6, cystatin E/M, cystatin M, cystatin M/E, Cystatin-6, Cystatin-E, cystatin-M, cysteine proteinase inhibitor |
| Host Species | Rat |
| Immunogen | Mouse recombinant Cystatin E/M/CST6 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?