Learn More
Cystatin E/M/CST6 Antibody (RM0217-6F49), CoraFluor™ 1, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP2-12252CL1
Product Code. 30146771
  • £771.00 / 0.10mL
Description
Cystatin E/M/CST6 Monoclonal antibody specifically detects Cystatin E/M/CST6 in Mouse samples. It is validated for Western Blot,Neutralization
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cystatin E/M/CST6
Monoclonal
CoraFluor 1
cystatin 6, cystatin E/M, cystatin M, cystatin M/E, Cystatin-6, Cystatin-E, cystatin-M, cysteine proteinase inhibitor
Mouse recombinant Cystatin E/M/CST6
0.1 mL
Cancer
1474
Store at 4°C in the dark. Do not freeze.
IgG2
Western Blot, Neutralization
RM0217-6F49
PBS
Rat
Protein A or G purified
RUO
Primary
Mouse
Purified