missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin E/M/CST6 Antibody (037), Janelia Fluor™ 549, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Cystatin E/M/CST6 Monoclonal antibody specifically detects Cystatin E/M/CST6 in Mouse samples. It is validated for ELISA
Specifications
Specifications
| Conjugate | Janelia Fluor 549 |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?