missing translation for 'onlineSavingsMsg'
Learn More

Cystatin C Antibody (Cyst13), DyLight 350, Novus Biologicals™

Product Code. 18422775 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.2 mL
Unit Size:
0.20mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18422775 0.2 mL 0.20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18422775 Supplier Novus Biologicals Supplier No. NBP105763UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cystatin C Monoclonal antibody specifically detects Cystatin C in Human samples. It is validated for ELISA, Immunoassay, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cystatin C
Applications ELISA, Immunoassay, Sandwich ELISA
Classification Monoclonal
Clone Cyst13
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias ARMD11, bA218C14.4 (cystatin C), cystatin 3, cystatin C, cystatin C (amyloid angiopathy and cerebral hemorrhage), cystatin-3, cystatin-C, Gamma-trace, MGC117328, Neuroendocrine basic polypeptide, post-gamma-globulin
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with cystatin C purified from human urine.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Cancer, Epitope Tags, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1471
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.