missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin B/Stefin B Antibody (CPTC-CSTB-3), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08538AF750
This item is not returnable.
View return policy
Description
Cystatin B/Stefin B Monoclonal antibody specifically detects Cystatin B/Stefin B in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| Cystatin B/Stefin B | |
| Monoclonal | |
| Alexa Fluor 750 | |
| CPI-B, CST6cystatin B (liver thiol proteinase inhibitor)10STFBcystatin-B, cystatin B (stefin B), EPM1, Liver thiol proteinase inhibitor, PME, stefin B, stefin-B | |
| Recombinant full-length human Cystatin B/Stefin B protein (Uniprot: P04080) | |
| 100 μg | |
| Cancer | |
| 1476 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| CPTC-CSTB-3 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction