missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin B Recombinant Rabbit Monoclonal Antibody (11H4)

Cystatin B Recombinant Rabbit Monoclonal Antibody (11H4)
Brand: Invitrogen MA550056
This item is not returnable.
View return policy
Description
Cystatin B Recombinant Monoclonal Antibody for ICC/IF, ELISA
Specifications
| Cystatin B | |
| Recombinant Monoclonal | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P04080 | |
| CSTB | |
| A synthesized peptide derived from human CSTB. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Immunocytochemistry | |
| 11H4 | |
| Unconjugated | |
| CSTB | |
| AA960480; CPI-B; Cst6; CSTB; Cyb; cystatin B; cystatin B (stefin B); cystatin B protein; cystatin beta; cystatin-B; Cystatin-beta; Epm1; EPM1A; Liver thiol proteinase inhibitor; LOC101123010; PME; stefin B; Stefin-B; stefin-C; Stfb; ULD; Unknown (protein for MGC:134609) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1476 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction