missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cystatin B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596258
This item is not returnable.
View return policy
Description
Immunogen sequence: MMCGAPSATQ PATAETQHIA DQVRSQLEEK ENKKFPVFKA VSFKSQVVAG TNYFIKVHVG DEDFVHLRVF QSLPHENKPL TLSNYQTNKA KHDELTYF; Positive Samples: HepG2, SW480, DU145, A-549, A-431, MCF7; Cellular Location: Cytoplasm, Nucleus.
Cystatin A (also designated STF1, STFA, stefin A or cystatin AS) and cystatin B (also designated PME, CST6, STFB, CPI-B, stefin B and liver thiol proteinase inhibitor) are thiol protease inhibitors that form complexes with Papain and the cathepsins B, H and L. Cystatin A, a cytoplasmic protein, is one of the precursor proteins of the cornified cell envelope in keratinocytes and plays a role in epidermal development and maintenance. Cystatin B protects against intracellular proteases leaking out of lysosomes and is primarily expressed in heart, liver and kidney.
Specifications
| Cystatin B | |
| Polyclonal | |
| Unconjugated | |
| CSTB | |
| AA960480; CPI-B; Cst6; CSTB; Cyb; cystatin B; cystatin B (stefin B); cystatin B protein; cystatin beta; cystatin-B; Cystatin-beta; Epm1; EPM1A; Liver thiol proteinase inhibitor; LOC101123010; PME; stefin B; Stefin-B; stefin-C; Stfb; ULD; Unknown (protein for MGC:134609) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1476, 25308 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.79 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P01041, P04080 | |
| CSTB | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-98 of human CSTB (NP_0000911). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction