Learn More
Invitrogen™ Cystatin B Polyclonal Antibody
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  Invitrogen™ PA546909
Product Code. 15537402
  • £443.00 / 100µg
Description
In Western blots, less than 2% cross-reactivity with recombinant human (rh) Cystatin A, recombinant mouse Cystatin B, rhCystatin C, rhCystatin D, rhCystatin E/M, rhCystatin S, rhCystatin SA, and rhCystatin SN is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Cystatin A (also designated STF1, STFA, stefin A or cystatin AS) and cystatin B (also designated PME, CST6, STFB, CPI-B, stefin B and liver thiol proteinase inhibitor) are thiol protease inhibitors that form complexes with Papain and the cathepsins B, H and L. Cystatin A, a cytoplasmic protein, is one of the precursor proteins of the cornified cell envelope in keratinocytes and plays a role in epidermal development and maintenance. Cystatin B protects against intracellular proteases leaking out of lysosomes and is primarily expressed in heart, liver and kidney.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cystatin B
Polyclonal
Unconjugated
CSTB
AA960480; CPI-B; Cst6; CSTB; Cyb; cystatin B; cystatin B (stefin B); cystatin B protein; cystatin beta; cystatin-B; Cystatin-beta; Epm1; EPM1A; Liver thiol proteinase inhibitor; LOC101123010; PME; stefin B; Stefin-B; stefin-C; Stfb; ULD; Unknown (protein for MGC:134609)
Goat
Antigen affinity chromatography
RUO
1476, 25308
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Immunohistochemistry, Immunoprecipitation, Western Blot
0.2 mg/mL
PBS with 5% trehalose and No Preservative
P01041, P04080
CSTB
E. coli-derived recombinant human Cystatin B Met2-Phe98.
100 μg
Primary
Human, Rat