missing translation for 'onlineSavingsMsg'
Learn More

Cystatin B Mouse anti-Human, Clone: 2G9B8, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66812-1-IG-20UL

 View more versions of this product

Product Code. 16829035

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Cystatin A (also designated STF1, STFA, stefin A or cystatin AS) and cystatin B (also designated PME, CST6, STFB, CPI-B, stefin B and liver thiol proteinase inhibitor) are thiol protease inhibitors that form complexes with Papain and the cathepsins B, H and L. Cystatin A, a cytoplasmic protein, is one of the precursor proteins of the cornified cell envelope in keratinocytes and plays a role in epidermal development and maintece. Cystatin B protects against intracellular proteases leaking out of lysosomes and is primarily expressed in heart, liver and kidney.
Specifications

Specifications

Cystatin B
Monoclonal
1.5 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P04080
CSTB
Cystatin B Fusion Protein Ag21291
20 μL
Primary
Human
Antibody
IgG1
Western Blot
2G9B8
Unconjugated
CSTB
CPI B, CST6, CSTB, Cystatin B, cystatin B (stefin B), EPM1, PME, Stefin B, STFB
Mouse
Protein G
RUO
1476
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.