missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cystatin B Monoclonal Antibody (C6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA550854
This item is not returnable.
View return policy
Description
Cystatin A (also designated STF1, STFA, stefin A or cystatin AS) and cystatin B (also designated PME, CST6, STFB, CPI-B, stefin B and liver thiol proteinase inhibitor) are thiol protease inhibitors that form complexes with Papain and the cathepsins B, H and L. Cystatin A, a cytoplasmic protein, is one of the precursor proteins of the cornified cell envelope in keratinocytes and plays a role in epidermal development and maintenance. Cystatin B protects against intracellular proteases leaking out of lysosomes and is primarily expressed in heart, liver and kidney.
Specifications
| Cystatin B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| P01041, Q62426 | |
| CSTB | |
| Recombinant Rat CSTB protein, Met2-Phe98 (Accession #P01041). | |
| 20 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG2b κ |
| Western Blot | |
| C6 | |
| Unconjugated | |
| CSTB | |
| AA960480; CPI-B; Cst6; CSTB; Cyb; cystatin B; cystatin B (stefin B); cystatin B protein; cystatin beta; cystatin-B; Cystatin-beta; Epm1; EPM1A; Liver thiol proteinase inhibitor; LOC101123010; PME; stefin B; Stefin-B; stefin-C; Stfb; ULD; Unknown (protein for MGC:134609) | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 13014, 25308 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction