missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin A, Mouse anti-Human, Clone: CSTA/2882, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79747-20UG
This item is not returnable.
View return policy
Description
Cystatin A Monoclonal specifically detects Cystatin A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Cystatin A | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| P01040, P01040 | |
| CSTA | |
| Recombinant fragment of human Cystatin A protein (around aa 1-98) (exact sequence is proprietary) | |
| 20 μg | |
| Cancer, Tumor Suppressors | |
| 1475 | |
| Store at 4°C. | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| CSTA/2882 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| cystatin A (stefin A), cystatin AS, cystatin-A, STF1Cystatin-AS, STFAStefin-A | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction