missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin A Antibody (SR1147), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22275-50ul
This item is not returnable.
View return policy
Description
Cystatin A Monoclonal antibody specifically detects Cystatin A in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry
Specifications
| Cystatin A | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry | |
| SR1147 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50 | |
| cystatin A (stefin A), cystatin AS, cystatin-A, STF1Cystatin-AS, STFAStefin-A | |
| A synthesized peptide derived from human Cystatin A (Uniprot #: P01040) | |
| 50 μg | |
| Cancer, Tumor Suppressors | |
| 1475 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu