missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cystatin 9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556080
This item is not returnable.
View return policy
Description
Immunogen sequence: GGNNKIVQDP MFFATVEFAL NTFNVQSKEE HAYRLLRVLS SWREDSMDRK WRGKMVFSMN LQLRQTVCRK FEDDIDNCPF QESLELNNVR QGISFPQVHS CGCCMGCGV Highest antigen sequence identity to the following orthologs: Mouse - 39%, Rat - 39%.
The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins and the kininogens. The type 2 cystatin proteins are a class of cysteine proteinase inhibitors found in a variety of human fluids and secretions, where they appear to provide protective functions. The cystatin locus on chromosome 20 contains the majority of the type 2 cystatin genes and pseudogenes. This gene is located in the cystatin locus and encodes a secreted protein that may play a role in hematopoietic differentiation or inflammation.
Specifications
| Cystatin 9 | |
| Polyclonal | |
| Unconjugated | |
| CST9 | |
| CLM; CST9; CTES7A; cystatin 9; cystatin 9 (testatin); cystatin-9; Cystatin-like molecule; testatin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 128822 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5W186 | |
| CST9 | |
| Recombinant protein corresponding to Human Cystatin 9. Recombinant protein control fragment (Product #RP-88587). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction