missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cystatin 2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA593068

Product Code. 16518371

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: AWSPQEEDRI IEGGIYDADL NDERVQRALH FVISEYNKAT EDEYYRRLLR VLRAREQIVG GVNYFFDIEV GRTICTKSQP NLDTCAFHEQ PELQKKQLCS FQIYEVPWED RMSLVNSRCQ EA; Positive Samples: Mouse heart.

The cystatin superfamily is a well-established family of cysteine protease inhibitors. Cystatins A and B (type 1) are mainly intracellular; cystatins C, D, E/M, F, G, S, SN and SA cystatins are extracellular (type 2); and the kininogens are type 3 cystatins which are intravascular proteins. All true cystatins inhibit cysteine peptidases of the papain family, such as cathepsins, and some also inhibit legumain family enzymes. Cystatin SA, cystatin S and cystatin SN are found primarily in saliva. Cystatin S and SN can also be expressed in tears, urine and seminal fluid. Cystatin C is a related protein which is expressed in brain, thymus, ovary, epididymis and vas deferens. Cystatin D protects against proteinases in the oral cavity, while Cystatin E/M and F moderate the inhibition of cathepsin proteins. The fetuins, part of the cystatin superfamily, are secretable proteins that influence osteogenesis and bone resorption, regulation of the insulin and hepatocyte growth factor receptors and the response to systemic inflammation. High molecular weight kininogen (Kininogen HC) and low molecular weight kininogen (Kininogen LC) have varied roles, though they both inhibit the thrombin- and plasmin-induced aggregation of thrombocytes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cystatin 2
Polyclonal
Unconjugated
CST2
CST2; cystatin 2; cystatin S5; cystatin SA; cystatin-2; Cystatin-S5; cystatin-SA; cysteine-proteinase inhibitor; salivary cysteine (thiol) protease inhibitor
Rabbit
Affinity chromatography
RUO
1470
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.51 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P09228
CST2
Recombinant fusion protein containing a sequence corresponding to amino acids 20-141 of human CST2 (NP_0013131).
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.