missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYSLTR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575328
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Receptor for cysteinyl leukotrienes. The response is mediated via a G-protein that activates a phosphatidylinositol-calcium second messenger system. Stimulation by BAY u9773, a partial agonist, induces specific contractions of pulmonary veins and might also have an indirect role in the relaxation of the pulmonary vascular endothelium. The rank order of affinities for the leukotrienes is LTC4 = LTD4 >> LTE4.
Specifications
| CYSLTR2 | |
| Polyclonal | |
| Unconjugated | |
| CYSLTR2 | |
| 2300001H05Rik; Cltr2; CysLT(2); Cyslt2; CysLT-2; CYSLT2R; CYSLTR2; cysteinyl leukotriene CysLT2 receptor; cysteinyl leukotriene receptor 2; GPCR21; G-protein coupled receptor GPCR21; G-protein coupled receptor HG57; HG57; hGPCR21; HPN321; KPG_011; PSEC0146; RSBPT32 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 170926, 57105, 70086 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q920A1, Q924T9, Q9NS75 | |
| CYSLTR2 | |
| Synthetic peptide corresponding to amino acids 290-336 of Human CysLTR2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction