missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYRPTIC/CFC1 Antibody (MM0205-8D12), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12209MFV610
This item is not returnable.
View return policy
Description
CYRPTIC/CFC1 Monoclonal antibody specifically detects CYRPTIC/CFC1 in Human samples. It is validated for Western Blot
Specifications
| CYRPTIC/CFC1 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 55997 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot | |
| MM0205-8D12 | |
| Western Blot | |
| CFC1B, cripto, FRL-1, cryptic family 1, Cryptic family protein 1, cryptic protein, CRYPTICFLJ77897, HTX2MGC133213 | |
| Human recombinant CYRPTIC/CFC1 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction