missing translation for 'onlineSavingsMsg'
Learn More

Cyrene, TRC

Product Code. 30363863
Change view
Click to view available options
Quantity:
50 mL
100 mL
250 mL
Unit Size:
100mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30363863 250 mL 250mL
30353802 100 mL 100mL
30357915 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30363863 Supplier Toronto Research Chemicals Supplier No. TRCC995413250ML
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53716-82-8
Molecular Formula C6H8O3
Molecular Weight (g/mol) 128.13
Synonym (1S,5R)-6,8-Dioxabicyclo[3.2.1]octan-4-one, (5R)-6,8-Dioxabicyclo[3.2.1]octan-4-one, Cyrene, Dihydrolevoglucosenone
IUPAC Name (1S,5R)-6,8-dioxabicyclo[3.2.1]octan-4-one
SMILES O=C1CC[C@H]2CO[C@@H]1O2

Specifications

Quantity 250 mL
InChI Formula InChI=1S/C6H8O3/c7-5-2-1-4-3-8-6(5)9-4/h4,6H,1-3H2/t4-,6+/m0/s1
Formula Weight 128.0473
Chemical Name or Material Cyrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.