Learn More
Novus Biologicals™ Cyr61/CCN1 Antibody (Roche patent anti-CCN1) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-27957-100ug
 View more versions of this product
Product Code. 30047622
  • £257.00 / 100µg
Description
Cyr61/CCN1 Monoclonal antibody specifically detects Cyr61/CCN1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cyr61/CCN1
Monoclonal
Unconjugated
O00622
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Roche patent anti-CCN1
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
CCN family member 1, CCN1IGF-binding protein 10, Cysteine-rich angiogenic inducer 61, cysteine-rich, angiogenic inducer, 61, GIG1cysteine-rich heparin-binding protein 61, IBP-10, IGFBP-10, IGFBP10cysteine-rich, anigogenic inducer, 61, Insulin-like growth factor-binding protein 10, protein CYR61, Protein GIG1
CYR61/ CCN1
100 μg
Biologically Active Proteins
3491
Human
Purified