missing translation for 'onlineSavingsMsg'
Learn More

Cyr61/CCN1 Antibody (365108) [CoraFluor™ 1], Novus Biologicals™

Product Code. 30733285 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733285 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733285 Supplier Novus Biologicals Supplier No. FAB4055CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cyr61/CCN1 Monoclonal antibody specifically detects Cyr61/CCN1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyr61/CCN1
Applications Western Blot
Classification Monoclonal
Clone 365108
Conjugate CoraFluor 1
Formulation PBS
Gene Alias CCN family member 1, CCN1IGF-binding protein 10, Cysteine-rich angiogenic inducer 61, cysteine-rich, angiogenic inducer, 61, GIG1cysteine-rich heparin-binding protein 61, IBP-10, IGFBP-10, IGFBP10cysteine-rich, anigogenic inducer, 61, Insulin-like growth factor-binding protein 10, protein CYR61, Protein GIG1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Cyr61, Ala22-Asp381, Accession # O00622
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 3491
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.