missing translation for 'onlineSavingsMsg'
Learn More

Cyproterone Acetate, Mikromol™

Product Code. 17909599
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 17909599

Brand: Mikromol MM0215.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Cyproterone Acetate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyproterone Acetate
CAS 427-51-0
Analyte or Component Names Cyproterone Acetate
Shipping Condition Room Temperature
Molecular Formula C24 H29 Cl O4
Certifications/Compliance ISO 17034
Synonym Cyproterone Acetate, Cyclopropa[1,2]cyclopenta[a]phenanthrene, 3'H-cyclopropa[1,2]pregna-1,4,6-triene-3,20-dione deriv., 3'H-Cyclopropa[1,2]pregna-1,4,6-triene-3,20-dione, Androcur, CPA, Cyprostat, Cyproterone 17-O-acetate, Cyproterone acetate, Cyproviron, NSC 81430, SH 714, (1alpha,2alpha)-17-(Acetyloxy)-6-chloro-1,2-dihydro-3'H-cyclopropa[1,2]pregna-1,4,6-triene-3,20-dione, 6-Chloro-1alpha,2alpha-dihydro-17-hydroxy-3'H-cyclopropa[1,2]pregna-1,4,6-triene-3,20-dione acetate, 6-Chloro-17-hydroxy-1alpha,2alpha-methylenepregna-4,6-diene-3,20-dione acetate, 1,2alpha-Methylene-6-chloro-17alpha-acetoxy-4,6-pregnadiene-3,20-dione, 1,2alpha-Methylene-6-chloro-pregna-4,6-diene-3,20-dione 17alpha-acetate, 1,2alpha-Methylene-6-chloro-Delta4,6-pregnadien-17alpha-ol-3,20-dione acetate, 17alpha-Acetoxy-6-chloro-1alpha,2alpha-methylenepregna-4,6-diene-3,20-dione, 6-Chloro-1,2alpha-methylene-17alpha-hydroxy-Delta6-progesterone acetate, 6-Chloro-1,2alpha-methylene-6-dehydro-17alpha-hydroxyprogesterone acetate, Cyproterone 17alpha-acetate
InChI Formula InChI=1S/C24H29ClO4/c1-12(26)24(29-13(2)27)8-6-16-14-10-20(25)19-11-21(28)15-9-18(15)23(19,4)17(14)5-7-22(16,24)3/h10-11,14-18H,5-9H2,1-4H3/t14-,15+,16-,17-,18-,22-,23-,24-/m0/s1
SMILES CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C=C(Cl)C4=CC(=O)[C@@H]5C[C@@H]5[C@]4(C)[C@H]3CC[C@]12C)C(=O)C
IUPAC Name (1α,2α)-17-(Acetyloxy)-6-chloro-1,2-dihydro-3'H-cyclopropa[1,2]pregna-1,4,6-triene-3,20-dione
Molecular Weight (g/mol) 416.94
Quantity 250 mg
Formula Weight 416.1754 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.