missing translation for 'onlineSavingsMsg'
Learn More

Cyproterone acetate, MedChemExpress

Product Code. 16481676
Encompass_Preferred
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
This item is not returnable. View return policy

Product Code. 16481676

Brand: Medchem Express HY13604

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cyproterone acetate is an anti-androgen (IC50=7.1 nM) and progestogen synthetic steroid. Cyproterone acetate has affinity with progesteron and with glucocorticoidal receptors.

Cyproterone acetate is an anti-androgen (IC50=7.1 nM) and progestogen synthetic steroid. Cyproterone acetate has affinity with progesteron and with glucocorticoidal receptors.

Specifications

Chemical Name or Material Cyproterone acetate
Physical Form Solid
CAS 427-51-0
Color Off-White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C23H27ClO4
Solubility Information DMSO : 33.33 mg/mL (82.72 mM; Need ultrasonic) ∣Ethanol : 20 mg/mL (49.64 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.25 mM; Need ultrasonic)
SMILES C[C@@]12C(C(Cl)=C[C@]3([H])[C@]2([H])C[C@@]4(C)[C@@]3([H])CC[C@@]4(C(C)=O)OC(C)=O)=CC([C@H]5[C@@H]1C5)=O
Molecular Weight (g/mol) 402.91
Formula Weight 402.91
Percent Purity 99.93%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.